missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Methylpyrazin-2-yl)ethan-1-one, 97%, Thermo Scientific™

Product Code. 10295884
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10295884 10 g 10g
10255934 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10295884 Supplier Thermo Scientific Maybridge Supplier No. SB01015EA
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 23787-80-6
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00014612
InChI Key QUNOTZOHYZZWKQ-UHFFFAOYSA-N
Synonym 2-acetyl-3-methylpyrazine, 1-3-methylpyrazin-2-yl ethanone, 1-3-methylpyrazin-2-yl ethan-1-one, ethanone, 1-3-methylpyrazinyl, 2-methyl-3-acetylpyrazine, unii-7u5tew2y2b, 2-acetyl-3-methyl pyrazine, 1-3-methylpyrazinyl ethan-1-one, 3-acetyl-2-methylpyrazine, ethanone, 1-3-methyl-2-pyrazinyl
PubChem CID 32093
IUPAC Name 1-(3-methylpyrazin-2-yl)ethanone
SMILES CC(=O)C1=NC=CN=C1C

Specifications

CAS Min % 97.0
Density 1.1100g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 136.15
Percent Purity 97%
Chemical Name or Material 1-(3-Methylpyrazin-2-yl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.