missing translation for 'onlineSavingsMsg'
Learn More

1,3-Phenylenediamine, TRC

Product Code. 30424189
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30424189 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30424189

Brand: Toronto Research Chemicals TRCP3198355G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Phenylenediamine
CAS 108-45-2
Molecular Formula C6 H8 N2
Quantity 5 g
Synonym 1,3-Benzenediamine, m-Phenylenediamine (8CI), 1,3-Diaminobenzene, 1,3-Diaminophenylene, 1,3-Phenylenediamine, 3-Aminoaniline, C.I. Developer 11, Developer C, Developer H, Developer M, Direct Brown BR, Direct Brown GG, MPDA, Maleid F, NSC 4776, RT 30H, m-Aminoaniline, m-Benzenediamine, m-Diaminobenzene
InChI Formula InChI=1S/C6H8N2/c7-5-2-1-3-6(8)4-5/h1-4H,7-8H2
SMILES Nc1cccc(N)c1
IUPAC Name benzene-1,3-diamine
Molecular Weight (g/mol) 108.14
Formula Weight 108.0687
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.