missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Propane-d6-diamine 2HCl, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1,3-Diaminopropane-D6 Dihydrochloride |
| CAS | 65898-86-4 |
| Molecular Formula | C3 D6 H4 N2 . 2 Cl H |
| Quantity | 0.25 g |
| Synonym | 1,3-Propane-1,1,2,2,3,3-d6-diamine, dihydrochloride (9 CI), 1,1,2,2,3,3-Hexadeuteriopropane-1,3-diamine dihydrochloride, 1,3-Diaminopropane-d6 dihydrochloride, 1,3-Diaminopropane-D6 dihydrochloride, Trimethylenediamine-d6 dihydrochloride, 1,3-Propane-d6-diamine dihydrochloride |
| InChI Formula | InChI=1 S/C3H10N2.2 ClH/c4-2-1-3-5;;/h1-5H2;2*1 H/i1D2,2D2,3D2;; |
| SMILES | Cl.Cl.[2 H]C([2 H])(N)C([2 H])([2 H])C([2 H])([2 H])N |
| IUPAC Name | 1,1,2,2,3,3-hexadeuteriopropane-1,3-diamine;dihydrochloride |
| Molecular Weight (g/mol) | 153.0838 |
| Formula Weight | 152.0754 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction