missing translation for 'onlineSavingsMsg'
Learn More

1,3-Propanediol di-p-toluenesulfonate, 98%, Thermo Scientific Chemicals

Product Code. 11414547
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414547 25 g 25g
11404547 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414547 Supplier Thermo Scientific Alfa Aesar Supplier No. B21084.14
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5469-66-9
Molecular Formula C17H20O6S2
Molecular Weight (g/mol) 384.46
MDL Number MFCD00013392
InChI Key ODVREDGIWSDQFD-UHFFFAOYSA-N
Synonym 1,3-bis tosyloxy propane, propane-1,3-diyl bis 4-methylbenzenesulfonate, 1,3-propanediol di-p-toluenesulfonate, 1,3-propanediol di-p-tosylate, 1,3-propanediol-di-4-tosylate, 3-4-methylbenzenesulfonyl oxy propyl 4-methylbenzene-1-sulfonate, 3-4-methylbenzenesulfonyl oxy propyl 4-methylbenzenesulfonate, propane-1,3-di-tos, acmc-209liv, 1,3-propanediol ditosylate
PubChem CID 230370
IUPAC Name 3-(4-methylphenyl)sulfonyloxypropyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCCOS(=O)(=O)C1=CC=C(C)C=C1

Specifications

Melting Point 90°C to 92°C
Quantity 25 g
Formula Weight 384.47
Percent Purity 98%
Chemical Name or Material 1,3-Propanediol di-p-toluenesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.