missing translation for 'onlineSavingsMsg'
Learn More

1,3-Propanediol, TRC

Product Code. 30137596
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137596 50 g 50g
30133116 10 g 10g
30007633 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137596 Supplier Toronto Research Chemicals Supplier No. TRCP76032050G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-63-2
Molecular Formula C3 H8 O2
Molecular Weight (g/mol) 76.09
Synonym 1,3-Propanediol, 1,3-Dihydroxypropane, 1,3-Propylene glycol, 1,3-Propylenediol, 2-Deoxyglycerol, Bio-PDO, NSC 65426, PG, Susterra, Trimethylene glycol, Zemea, Zemea propanediol, β-Propylene glycol, ω-Propanediol
IUPAC Name propane-1,3-diol
SMILES OCCCO

Specifications

Quantity 50 g
InChI Formula InChI=1S/C3H8O2/c4-2-1-3-5/h4-5H,1-3H2
Formula Weight 76.0524
Chemical Name or Material 1,3-Propanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.