missing translation for 'onlineSavingsMsg'
Learn More

1,3-Propanedithiol, 97%

Product Code. 11482308
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482308 100 g 100g
11462308 5 g 5g
11472308 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482308 Supplier Thermo Scientific Alfa Aesar Supplier No. A15261.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 109-80-8 | C3H8S2 | 108.217 g/mol

1,3-Propanedithiol is used as a reagent in the preparation of thioketals and thioacetals. It acts as a flavoring agent. It is used as a precursor in the synthesis of cyclic dithioacetal (1,3-dithiane) derivatives of carbonyl compounds. It is involved in the preparation of diiron propanedithiolate hexacarbonyl by reacting reaction with triiron dodecacarbonyl. Further, it is used for the protection of aldehydes and ketones through their reversible formation of dithianes. In addition to this, it reacts with metal ions to form chelate rings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Propanedithiol is used as a reagent in the preparation of thioketals and thioacetals. It acts as a flavoring agent. It is used as a precursor in the synthesis of cyclic dithioacetal (1,3-dithiane) derivatives of carbonyl compounds. It is involved in the preparation of diiron propanedithiolate hexacarbonyl by reacting reaction with triiron dodecacarbonyl. Further, it is used for the protection of aldehydes and ketones through their reversible formation of dithianes. In addition to this, it reacts with metal ions to form chelate rings.

Solubility
Slightly soluble in water. Miscible with alcohol, ether, chloroform, etherMiscible with alcohol, ether, chloroform and ether. Slightly miscible with water.

Notes
Store in a cool place. Incompatible with bases, oxidizing agents, reducing agents and alkali metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-80-8
Molecular Formula C3H8S2
Molecular Weight (g/mol) 108.217
MDL Number MFCD00004904
InChI Key ZJLMKPKYJBQJNH-UHFFFAOYSA-N
Synonym 1,3-propanedithiol, 1,3-dimercaptopropane, trimethylene dimercaptan, trimethylenedithiol, dithiotrimethyleneglycol, trimethylenedithioglycol, 1,3-propanedimercaptan, unii-r4luj82u52, fema no. 3588, 1,3-propane dithiol
PubChem CID 8013
ChEBI CHEBI:44864
IUPAC Name propane-1,3-dithiol
SMILES C(CS)CS

Specifications

Melting Point -79°C
Density 1.077
Boiling Point 168°C to 170°C
Flash Point 63°C (145°F)
Odor Stench
Refractive Index 1.54
Quantity 100 g
UN Number UN3334
Beilstein 1071197
Merck Index 14,7801
Solubility Information Slightly soluble in water; Miscible with alcohol,ether,chloroform,etherMiscible with alcohol,ether,chloroform and ether; Slightly miscible with water.
Formula Weight 108.23
Percent Purity 97%
Chemical Name or Material 1,3-Propanedithiol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.