missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Pyridyl)ethylamine, 96%

Product Code. 15443357
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15443357 250 mg 250mg
11351979 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15443357 Supplier Thermo Scientific Alfa Aesar Supplier No. H26047.MD
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56129-55-6 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56129-55-6
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD01691641,MFCD09256803
InChI Key IQVQNBXPYJGNEA-UHFFFAOYNA-N
Synonym 1-pyridin-3-yl-ethylamine, 1-3-pyridyl ethylamine, 1-pyridin-3-yl ethanamine, 3-1-aminoethyl pyridine, 1-3-pyridinyl ethanamine, 1-pyridin-3-yl ethan-1-amine, alpha-methyl-3-pyridinemethylamine, 3-pyridinemethylamine, alpha-methyl, 1-pyridin-3-ylethyl amine, 1-3-pyridyl ethanamine
PubChem CID 2771688
IUPAC Name 1-pyridin-3-ylethanamine
SMILES CC(N)C1=CC=CN=C1

Specifications

Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 122.17
Percent Purity 96%
Chemical Name or Material 1-(3-Pyridyl)ethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.