missing translation for 'onlineSavingsMsg'
Learn More

1,3-Thiazol-5-ylmethylamine hydrochloride, 97%, Thermo Scientific™

Product Code. 10586595
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10586595 5 g Amber Glass Bottle 5g
10342145 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10586595 Supplier Thermo Scientific Maybridge Supplier No. MO08602DE
 Restricted Item
Estimated Shipment: 03-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 131052-46-5
Molecular Formula C4H7ClN2S
Molecular Weight (g/mol) 150.62
MDL Number MFCD06738808
InChI Key WZJFBMHEYMPACF-UHFFFAOYSA-N
Synonym thiazol-5-ylmethanamine hydrochloride, 5-aminomethylthiazole hcl, c-thiazol-5-yl-methylamine hydrochloride, 1,3-thiazol-5-ylmethanamine hydrochloride, thiazole-5-methylamine hcl, 5-aminomethylthiazole hydrochloride, 5-thiazol-methylamine hydrochloride, thiazol-5-yl methanamine hydrochloride, 1,3-thiazol-5-ylmethylamine hydrochloride, 5-thiazolemethanamine hydrochloride
PubChem CID 17750968
SMILES [H+].[Cl-].NCC1=CN=CS1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
IUPAC Name hydrogen 1-(1,3-thiazol-5-yl)methanamine chloride
Formula Weight 187.09
Percent Purity 97%
Chemical Name or Material 1, 3-Thiazol-5-ylmethylamine di-hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.