missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(4-Morpholinyl)phenyl]guanidine, 98%

Product Code. 15436577
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436577 1 g 1g
15446577 5 g 5g
15456577 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15436577 Supplier Thermo Scientific Alfa Aesar Supplier No. H33726.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 247234-41-9 | C11H16N4O | 220.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 247234-41-9
Molecular Formula C11H16N4O
Molecular Weight (g/mol) 220.28
MDL Number MFCD11986905
InChI Key ZVZJREQBRCRGLM-UHFFFAOYSA-N
Synonym 1-4-morpholinophenyl guanidine, n-4-morpholin-4-ylphenyl guanidine, 4-morpholinophenylguanidine, 4-morpholino phenylguanidine, n-4-morpholin-4-yl-phenyl-guanidine, n-4-morpholin-4-yl phenyl guanidine, 1-4-4-morpholinyl phenyl guanidine, 1-4-morpholin-4-yl phenyl guanidine, 2-4-morpholin-4-yl phenyl guanidine
PubChem CID 10176830
IUPAC Name 2-(4-morpholin-4-ylphenyl)guanidine
SMILES NC(N)=NC1=CC=C(C=C1)N1CCOCC1

Specifications

Melting Point ∼220°C (decomposition)
Density 1.309
Quantity 1 g
Formula Weight 220.27
Percent Purity 98%
Chemical Name or Material 1-[4-(4-Morpholinyl)phenyl]guanidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.