missing translation for 'onlineSavingsMsg'
Learn More

1,4,7-Triazonane, TRC

Product Code. 30457358
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457358 50 mg -
30455573 100 mg 100mg
30448941 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457358 Supplier Toronto Research Chemicals Supplier No. TRCT79527350MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4730-54-5
Molecular Formula C6H15N3
Molecular Weight (g/mol) 129.2
IUPAC Name 1,4,7-triazonane
SMILES C1CNCCNCCN1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H15N3/c1-2-8-5-6-9-4-3-7-1/h7-9H,1-6H2
Formula Weight 129.1266
Chemical Name or Material 1,4,7-Triazonane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.