missing translation for 'onlineSavingsMsg'
Learn More

1,4,8,11-Tetraazatricyclo[9.3.1.1(4,8)]hexadecane

Product Code. 15467605
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467605 250 mg 250mg
15493517 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467605 Supplier Thermo Scientific Alfa Aesar Supplier No. H26674.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 75920-10-4 | C12H24N4 | 224.352 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75920-10-4
Molecular Formula C12H24N4
Molecular Weight (g/mol) 224.352
MDL Number MFCD09953472
InChI Key ZJCWEHQOOSNWTF-UHFFFAOYSA-N
Synonym 1,4,8,11-tetraazatricyclo 9.3.1.1 4,8 hexadecane, 1,4,8,11-tetraazatricyclo 9.3.1.14,8 hexadecane, 1,4,8,11-tetraazatricyclo 9.3.1.1?,? hexadecane, formaldehyde-cyclam, acmc-20ap3h, cyclam formaldehyde derivative, 1,4,8,11-tetraazatricyclohexadecane, 1,4,8,11-tetraazatricyclo 9.3.1.1∼4,8∼ hexadecane
PubChem CID 337907
SMILES C1CN2CCN3CCCN(C3)CCN(C1)C2

Specifications

Melting Point 83°C to 85°C
Quantity 250 mg
Sensitivity Hygroscopic
Formula Weight 224.35
Chemical Name or Material 1,4,8,11-Tetraazatricyclo[9.3.1.1(4,8)]hexadecane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.