missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Acetylpiperidino)ethan-1-one, 97%, Thermo Scientific™

Product Code. 10331473
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10331473 1 g Amber Glass Bottle 1g
10321473 250 mg Amber Glass Bottle 250mg
10230135 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10331473 Supplier Thermo Scientific Maybridge Supplier No. CD11879DA
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 162368-01-6
Molecular Formula C9H15NO2
Molecular Weight (g/mol) 169.22
MDL Number MFCD00106774
InChI Key VFZXZEJGLKOTBW-UHFFFAOYSA-N
Synonym 1-4-acetylpiperidino ethan-1-one, 1-1-acetylpiperidin-4-yl ethanone, 1,4-diacetylpiperidine, 1-4-acetylpiperidin-1-yl ethan-1-one, 1,1'-piperidine-1,4-diyl diethanone, 1-1-acetyl-piperidin-4-yl-ethanone, piperidine, 1,4-diacetyl-9ci, 1-4-acetylpiperidin-1-yl ethanone, ethanone,1-1-acetyl-4-piperidinyl, piperidine, 1,4-diacetyl
PubChem CID 2777250
IUPAC Name 1-(1-acetylpiperidin-4-yl)ethanone
SMILES CC(=O)C1CCN(CC1)C(C)=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 169.22
Percent Purity 97%
Chemical Name or Material 1-(4-Acetylpiperidino)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.