missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Amino-2-hydroxyphenyl)ethan-1-one, Tech., Thermo Scientific™

Product Code. 10748304
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10748304 1 g Amber Glass Bottle 1g
10644903 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10748304 Supplier Thermo Scientific Maybridge Supplier No. BTB10056DA
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2476-29-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00100636
InChI Key QQZFVONVJPXCSQ-UHFFFAOYSA-N
Synonym 1-4-amino-2-hydroxyphenyl ethanone, 1-4-amino-2-hydroxyphenyl ethan-1-one, 4'-amino-2'-hydroxyacetophenone, 1-4-amino-2-hydroxyphenyl ethane-1-one, ethanone, 1-4-amino-2-hydroxyphenyl, 1-4-amino-2-hydroxy-phenyl ethanone, 2-acetyl-5-aminophenol, intermediates-zcf02667, amino-2-hydroxyphenyl ethanone, 4-acetyl-3-hydroxyaniline
PubChem CID 459296
IUPAC Name 1-(4-amino-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(O)C=C(N)C=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 151.16
Percent Purity Tech.
Concentration or Composition (by Analyte or Components) Technical Grade
Chemical Name or Material 1-(4-Amino-2-hydroxyphenyl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.