missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Aminobenzyl)-1H-1,2,4-triazole, 98%, Thermo Scientific™

Product Code. 15482789
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
15482789 25 g 25g
15492789 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 15482789

Brand: Thermo Scientific Alfa Aesar H66620.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119192-10-8
Molecular Formula C9H10N4
Molecular Weight (g/mol) 174.207
MDL Number MFCD00973306
InChI Key ZGLQVRIVLWGDNA-UHFFFAOYSA-N
Synonym 4-1h-1,2,4-triazol-1-ylmethyl aniline, 4-1h-1,2,4-triazol-1-yl methyl aniline, 1-4-aminobenzyl-1,2,4-triazole, 4-1,2,4-triazol-1-ylmethyl aniline, 4-1h-1,2,4-triazol-1-yl-methyl benzenamine, 4-1,2,4 triazol-1-ylmethylphenylamine, benzenamine, 4-1h-1,2,4-triazol-1-ylmethyl, 4-1,2,4 triazol-1-ylmethyl-phenylamine, 4-1h-1,2,4-triazol-1-yl-methyl aniline, 4-1h-1,2,4-triazol-1-ylmethyl benzenamine
PubChem CID 821219
IUPAC Name 4-(1,2,4-triazol-1-ylmethyl)aniline
SMILES C1=CC(=CC=C1CN2C=NC=N2)N

Specifications

Melting Point 126°C to 130°C
Color White to Yellow
Quantity 25 g
UN Number UN2811
Beilstein 5934828
Formula Weight 174.21
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-(4-Aminobenzyl)-1H-1,2,4-triazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.