missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzenediboronic acid bis(pinacol) ester, 97%

Product Code. 15478157
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15478157 5 g 5g
15468157 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15478157 Supplier Thermo Scientific Alfa Aesar Supplier No. H51707.06
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 99770-93-1 | C18H28B2O4 | 330.038 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 99770-93-1
Molecular Formula C18H28B2O4
Molecular Weight (g/mol) 330.038
MDL Number MFCD08276852
InChI Key UOJCDDLTVQJPGH-UHFFFAOYSA-N
Synonym 1,4-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene, 1,4-benzenediboronic acid bis pinacol ester, 4,4,5,5-tetramethyl-2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3,2-dioxaborolane, 1,4-phenylenediboronic acid, pinacol ester, 2,2'-1,4-phenylene bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2,2'-1,4-phenylene bis 4,4,5,5-tetramethyl, 4,4,5,5-tetramethyl-2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3,2-dioxaborolane, 1,4-benzenediboronic acid dipinacol ester, amtb185, 1,4-phenylenediboronic acid pinacol ester
PubChem CID 10592495
IUPAC Name 4,4,5,5-tetramethyl-2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)B3OC(C(O3)(C)C)(C)C

Specifica

Quantity 5 g
Formula Weight 330.03
Percent Purity 97%
Chemical Name or Material 1,4-Benzenediboronic acid bis(pinacol) ester

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.