missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxan-2-carbonitrile, TRC

Product Code. 30335229
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30335229 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335229 Supplier Toronto Research Chemicals Supplier No. TRCB2035102G
 Restricted Item
Estimated Shipment: 10-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spezifikation

Chemical Name or Material 1,4-Benzodioxan-2-carbonitrile
CAS 1008-92-0
Molecular Formula C9H7NO2
Quantity 2 g
Synonym 2,3-Dihydro-1,4-benzodioxin-2-carbonitrile, 2-Cyano-1,4-benzodioxan.
InChI Formula InChI=1S/C9H7NO2/c10-5-7-6-11-8-3-1-2-4-9(8)12-7/h1-4,7H,6H2
SMILES N#CC1COc2ccccc2O1
IUPAC Name 2,3-dihydro-1,4-benzodioxine-3-carbonitrile
Molecular Weight (g/mol) 161.16
Formula Weight 161.0477
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.