missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxan-5-amine, 95%

Product Code. 10422750 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10422750 1 g Glass bottle 1g
10753132 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10422750 Supplier Thermo Scientific Acros Supplier No. 429370010
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 16081-45-1 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16081-45-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD03695459
InChI Key DMLRSJNZORFCBD-UHFFFAOYSA-N
Synonym 5-amino-1,4-benzodioxane, 2,3-dihydrobenzo b 1,4 dioxin-5-amine, 2,3-dihydro-benzo 1,4 dioxin-5-ylamine, 2,3-ethylenedioxyaniline, 1,4-benzodioxin-5-amine,2,3-dihydro, 1,4-benzodioxan-5-amine, 5-amino-1,4-benzodioxan, 5-amino-1,4-dibenzodioxane, 1,4-benzodioxin-5-amine, 2,3-dihydro, 5-amino-2,3-dihydro-1,4-benzodioxin
PubChem CID 11788387
IUPAC Name 2,3-dihydro-1,4-benzodioxin-5-amine
SMILES C1COC2=C(C=CC=C2O1)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow to Amber
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 151.17
Percent Purity 95%
Physical Form Beads
Chemical Name or Material 1, 4-Benzodioxan-5-amine
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Benzodioxan-5-amine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.