missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxane-6-boronic acid, 97%

Product Code. 11349156
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11349156 5 g 5g
11339156 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349156 Supplier Thermo Scientific Alfa Aesar Supplier No. L20296.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 164014-95-3 | C8H9BO4 | 179.966 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 164014-95-3
Molecular Formula C8H9BO4
Molecular Weight (g/mol) 179.966
MDL Number MFCD01009696
InChI Key SQDUGGGBJXULJR-UHFFFAOYSA-N
Synonym 1,4-benzodioxane-6-boronic acid, 2,3-dihydrobenzo b 1,4 dioxin-6-yl boronic acid, 3,4-ethylenedioxy benzeneboronic acid, 2,3-dihydro-1,4-benzodioxin-6-ylboranediol, boronic acid, 2,3-dihydro-1,4-benzodioxin-6-yl, 1,4-benzodioxan-6-boronic acid, 2,3-dihydro-1,4-benzodioxin-6-yl boronic acid, 2,3-dihydrobenzo b 1,4 dioxin-6-ylboronic acid, 1,4-benzodioxan-6-yl boronic acid
PubChem CID 2776178
IUPAC Name 2,3-dihydro-1,4-benzodioxin-6-ylboronic acid
SMILES B(C1=CC2=C(C=C1)OCCO2)(O)O

Spezifikation

Quantity 5 g
Beilstein 7478648
Formula Weight 179.97
Percent Purity 97%
Chemical Name or Material 1,4-Benzodioxane-6-boronic acid

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.