missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxane-6-carboxaldehyde, 99%, Thermo Scientific Chemicals

Product Code. 11452978
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452978 5 g 5g
11442978 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11452978 Supplier Thermo Scientific Alfa Aesar Supplier No. A18696.06
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 29668-44-8 | Molecular Formula: C9H8O3 | Molecular Weight: 164.16 g/mol

1,4-Benzodioxan-6-carboxaldehyde finds its uses as building block in the synthesis of tetrahydroisoquinolinones, in the preparation of benzofuran analog, a potential inhibitor of CAMP-specific phosphodiesterase type IV, in the synthesis of (5Z)-5-(2,3-dihydro-1,4-benzodioxan-6-ylmethylene)-1-methyl-2-thioxoimidazolidin-4-one. It was screened as a fragment for inhibitors of PDEA using enthalpy arrays and X-ray crystallography. 1,4-Benzodioxane-6-carboxaldehyde have also been used as an intermediate in the synthesis of a new class of Dopamine D3 and D4 receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Benzodioxan-6-carboxaldehyde finds its uses as building block in the synthesis of tetrahydroisoquinolinones, in the preparation of benzofuran analog, a potential inhibitor of CAMP-specific phosphodiesterase type IV, in the synthesis of (5Z)-5-(2,3-dihydro-1,4-benzodioxan-6-ylmethylene)-1-methyl-2-thioxoimidazolidin-4-one. It was screened as a fragment for inhibitors of PDEA using enthalpy arrays and X-ray crystallography. 1,4-Benzodioxane-6-carboxaldehyde have also been used as an intermediate in the synthesis of a new class of Dopamine D3 and D4 receptor antagonists.

Solubility
Solubility in methanol is almost transparent.

Notes
Store away from air, oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29668-44-8
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00010092
InChI Key CWKXDPPQCVWXAG-UHFFFAOYSA-N
Synonym 1,4-benzodioxan-6-carboxaldehyde, 1,4-benzodioxan-6-carbaldehyde, 2,3-dihydrobenzo b 1,4 dioxine-6-carbaldehyde, 1,4-benzodioxane-6-carboxaldehyde, 3,4-ethylenedioxybenzaldehyde, 1,4-benzodioxin-6-carboxaldehyde, 2,3-dihydro, 7-formyl-1,4-benzodioxane, 1,4-benzodioxane-6-aldehyde, 2,3-dihydro-1,4-benzodioxin-6-carbaldehyde, 1,4-benzodioxane-6-carbaldehyde
PubChem CID 248127
IUPAC Name 2,3-dihydro-1,4-benzodioxine-6-carbaldehyde
SMILES C1COC2=C(O1)C=CC(=C2)C=O

Specifications

Melting Point 48°C to 52°C
Boiling Point 104°C to 106°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 164.16
Percent Purity 99%
Chemical Name or Material 1,4-Benzodioxane-6-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.