missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(3-aminopropyl)piperazine, 98%

Product Code. 11313206
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313206 100 g 100g
11303206 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313206 Supplier Thermo Scientific Alfa Aesar Supplier No. L12508.22
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7209-38-3 | C10H24N4 | 200.33 g/mol

1,4-Bis(3-aminopropyl)piperazine was used to functionalize natural montmorillonite (M) and synthetic kanemite (K)1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Bis(3-aminopropyl)piperazine was used to functionalize natural montmorillonite (M) and synthetic kanemite (K)1.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7209-38-3
Molecular Formula C10H24N4
Molecular Weight (g/mol) 200.33
MDL Number MFCD00006161
InChI Key XUSNPFGLKGCWGN-UHFFFAOYSA-N
Synonym 1,4-bis 3-aminopropyl piperazine, 1,4-piperazinedipropanamine, 3,3'-piperazine-1,4-diyl bis propan-1-amine, bis aminopropyl piperazine, n,n'-bis 3-aminopropyl piperazine, unii-7gvx6c734b, bapp, ccris 8915, 1,4-di 3-aminopropyl piperazine, 3,3'-piperazine-1,4-diyldipropan-1-amine
PubChem CID 81629
IUPAC Name 3-[4-(3-aminopropyl)piperazin-1-yl]propan-1-amine
SMILES C1CN(CCN1CCCN)CCCN

Specifications

Melting Point 14°C
Density 0.973
Boiling Point 150°C to 152°C (2 mmHg)
Flash Point 162°C (323°F)
Refractive Index 1.5018
Quantity 100 g
UN Number UN2735
Beilstein 111747
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 200.33
Percent Purity 98%
Chemical Name or Material 1,4-Bis(3-aminopropyl)piperazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.