missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(phenylethynyl)benzene, 97%

Product Code. 15433927
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433927 25 g 25g
15413927 1 g 1g
15423927 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15433927 Supplier Thermo Scientific Alfa Aesar Supplier No. H30395.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1849-27-0 | C22H14 | 278.354 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1849-27-0
Molecular Formula C22H14
Molecular Weight (g/mol) 278.354
MDL Number MFCD00160829
InChI Key FPVSTPLZJLYNMB-UHFFFAOYSA-N
Synonym 1,4-bis phenylethynyl benzene, 1,4-bis 2-phenylethynyl benzene, benzene,1,4-bis 2-phenylethynyl, p-bis phenylethynyl benzene, 1,4-bis phenylethynyl benzene #, 1,4-bis 2-phenyl-ethynyl-benzene
PubChem CID 624226
IUPAC Name 1,4-bis(2-phenylethynyl)benzene
SMILES C1=CC=C(C=C1)C#CC2=CC=C(C=C2)C#CC3=CC=CC=C3

Specifications

Melting Point 176°C to 178°C
Quantity 25 g
Formula Weight 278.36
Percent Purity 97%
Chemical Name or Material 1,4-Bis(phenylethynyl)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.