missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromophenyl)pyrrolidine, 97%, Thermo Scientific™

Product Code. 10691265
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10691265 1 g 1g
10239942 250 mg 250mg
10356284 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10691265 Supplier Thermo Scientific Maybridge Supplier No. CC30110DA
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 22090-26-2
Molecular Formula C10H12BrN
Molecular Weight (g/mol) 226.117
InChI Key BVEGBJXZICCEQW-UHFFFAOYSA-N
Synonym 1-4-bromophenyl pyrrolidine, n-4-bromophenyl pyrrolidine, 1-4-bromophenyl-pyrrolidine, acmc-209frt, 4-pyrrolidinobromobenzene, 4-bromophenyl pyrrolidine, 1-4-bromo-phenyl-pyrrolidine, pyrrolidine,1-4-bromophenyl, 4-bromo-1-1-pyrrolidinyl benzene
PubChem CID 7016457
IUPAC Name 1-(4-bromophenyl)pyrrolidine
SMILES C1CCN(C1)C2=CC=C(C=C2)Br

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 226.12
Percent Purity 97%
Chemical Name or Material 1-(4-Bromophenyl)pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.