missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Butane-1,1,4,4-d4-diol, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1,4-Butanediol-1,1,4,4-D4 |
| CAS | 36684-44-3 |
| Molecular Formula | C4 D4 H6 O2 |
| Quantity | 0.1 g |
| Synonym | 1,4-Butane-1,1,4,4-d4-diol (9 CI), 1,1,4,4-Tetradeutero-1,4-butanediol, 1,4-Butanediol-1,1,4,4-d4, 1,4-Butanediol-1,1,4,4-D4, 1,4-Butanediol-d4 |
| InChI Formula | InChI=1 S/C4H10O2/c5-3-1-2-4-6/h5-6 H,1-4H2/i3D2,4D2 |
| SMILES | [2 H]C([2 H])(O)CCC([2 H])([2 H])O |
| IUPAC Name | 1,1,4,4-tetradeuteriobutane-1,4-diol |
| Molecular Weight (g/mol) | 94.1457 |
| Formula Weight | 94.0932 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction