missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butanediol, 99%

Product Code. 11382176
Change view
Click to view available options
Quantity:
250 g
1000 g
2500 g
Unit Size:
1000g
2500g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382176 250 g 250g
11456720 1000 g 1000g
11466720 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11382176 Supplier Thermo Scientific Alfa Aesar Supplier No. L03491.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 110-63-4 | C4H10O2 | 90.12 g/mol

1,4-Butanediol is commonly used as a solvent in the chemical industry to manufacture gamma-butyrolactone and elastic fibers like spandex. It is used as a cross-linking agent for thermoplastic urethanes, polyester plasticizers, paints and coatings. It undergoes dehydration in the presence of phosphoric acid yielded teterahydrofuran, which is an important solvent used for various applications. It acts an intermediate and is used to manufacture polytetramethylene ether glycol (PTMEG), polybutylene terephthalate (PBT) and polyurethane (PU). It finds application as an industrial cleaner and a glue remover.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Butanediol is commonly used as a solvent in the chemical industry to manufacture gamma-butyrolactone and elastic fibers like spandex. It is used as a cross-linking agent for thermoplastic urethanes, polyester plasticizers, paints and coatings. It undergoes dehydration in the presence of phosphoric acid yielded teterahydrofuran, which is an important solvent used for various applications. It acts an intermediate and is used to manufacture polytetramethylene ether glycol (PTMEG), polybutylene terephthalate (PBT) and polyurethane (PU). It finds application as an industrial cleaner and a glue remover.

Solubility
Miscible with water, ethanol, dimethyl sulfoxide and glycol ethers. Slightly miscible with diethyl ether. Immiscible with aliphatic and aromatic hydrocarbons.

Notes
Incompatible with strong oxidizing agents, acid chlorides, acid anhydrides and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-63-4
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00002968
InChI Key WERYXYBDKMZEQL-UHFFFAOYSA-N
Synonym 1,4-butanediol, 1,4-butylene glycol, tetramethylene glycol, 1,4-dihydroxybutane, 1,4-tetramethylene glycol, tetramethylene 1,4-diol, sucol b, diol 14b, agrisynth b1d, unii-7xoo2le6g3
PubChem CID 8064
ChEBI CHEBI:41189
IUPAC Name butane-1,4-diol
SMILES OCCCCO

Specifications

Melting Point 19°C to 20°C
Density 1.017
Boiling Point 229°C to 230°C
Flash Point 135°C (275°F)
Refractive Index 1.446
Quantity 250 g
Beilstein 1633445
Sensitivity Hygroscopic
Solubility Information Miscible with water,ethanol,dimethyl sulfoxide and glycol ethers. Slightly miscible with diethyl ether. Immiscible with aliphatic and aromatic hydrocarbons.
Formula Weight 90.12
Percent Purity 99%
Chemical Name or Material 1,4-Butanediol
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Butanediol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: 1,4-Butanediol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.