missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butanediol diglycidyl ether, 60%, technical, Thermo Scientific Chemicals

Product Code. 10235820 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10235820 500 g 500g
10105693 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10235820 Supplier Thermo Scientific Acros Supplier No. 154315000
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2425-79-8
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00005146
InChI Key SHKUUQIDMUMQQK-UHFFFAOYNA-N
Synonym 1,4-butanediol diglycidyl ether, 1,4-diglycidyloxybutane, 1,4-bis glycidyloxy butane, butanediol diglycidyl ether, araldit dy 026, 1,4-butane diglycidyl ether, grilonit rv 1806, 1,4-diglycidloxybutane, chs-rr2, 1,4-bis 2,3-epoxypropoxy butane
PubChem CID 17046
IUPAC Name 2-[4-(oxiran-2-ylmethoxy)butoxymethyl]oxirane
SMILES C(CCOCC1CO1)COCC1CO1

Specifications

CAS Min % 60
Color Yellow
Density 1.1000g/mL
Boiling Point 266.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 55% min. (GC)
Packaging Glass bottle
Refractive Index 1.4600 to 1.4650
Quantity 500 g
Specific Gravity 1.1
Solubility Information Solubility in water: 55,6g/L (20°C).
Viscosity <15 mPa.s (25°C)
Formula Weight 202.25
Percent Purity 60%
Grade Technical
Physical Form Liquid
Chemical Name or Material 1, 4-Butanediol diglycidyl ether, 60%
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Butanediol diglycidyl ether
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.