missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butanediol diglycidyl ether, 96%

Product Code. 11353276
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11353276 10 g 10g
11363276 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353276 Supplier Thermo Scientific Alfa Aesar Supplier No. L12632.09
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2425-79-8 | C10H18O4 | 202.25 g/mol

1,4-Butanediol diglycidyl ether is used for preparing amylose, xylan and hydroxyethyl-cellulose, to cross link polyethylenimine which also serves as a hole-blocking layer in organic solar cells, to cross link hyaluronic acid into hydrogels and for the activation of soluble dextran polymers. It is the most commonly used homobifunctional epoxide compound

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Butanediol diglycidyl ether is used for preparing amylose, xylan and hydroxyethyl-cellulose, to cross link polyethylenimine which also serves as a hole-blocking layer in organic solar cells, to cross link hyaluronic acid into hydrogels and for the activation of soluble dextran polymers. It is the most commonly used homobifunctional epoxide compound

Solubility
Soluble in water (55,6 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2425-79-8
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00005146
InChI Key SHKUUQIDMUMQQK-UHFFFAOYNA-N
Synonym 1,4-butanediol diglycidyl ether, 1,4-diglycidyloxybutane, 1,4-bis glycidyloxy butane, butanediol diglycidyl ether, araldit dy 026, 1,4-butane diglycidyl ether, grilonit rv 1806, 1,4-diglycidloxybutane, chs-rr2, 1,4-bis 2,3-epoxypropoxy butane
PubChem CID 17046
IUPAC Name 2-[4-(oxiran-2-ylmethoxy)butoxymethyl]oxirane
SMILES C(CCOCC1CO1)COCC1CO1

Specifications

Density 1.049
Boiling Point 157°C to 158°C (11 mmHg)
Flash Point 140°C (284°F)
Refractive Index 1.455
Quantity 10 g
Beilstein 115238
Sensitivity Hygroscopic
Solubility Information Soluble in water (55,6g/L at 20°C).
Formula Weight 202.25
Percent Purity 96%
Chemical Name or Material 1,4-Butanediol diglycidyl ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,4-Butanediol diglycidyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1,4-Butanediol diglycidyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.