missing translation for 'onlineSavingsMsg'
Learn More

1-(4-chlorophenethyl)-5-oxo-3-pyrrolidinecarboxylic acid, 97%, Thermo Scientific™

Product Code. 10743443
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10743443 1 g Amber Glass Bottle 1g
10753443 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10743443 Supplier Thermo Scientific Maybridge Supplier No. MO00129DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 368870-06-8
Molecular Formula C13H14ClNO3
Molecular Weight (g/mol) 267.709
MDL Number MFCD01330893
InChI Key IMVZABZPKCETQX-UHFFFAOYSA-N
Synonym 1-4-chlorophenethyl-5-oxopyrrolidine-3-carboxylic acid, 1-2-4-chlorophenyl ethyl-5-oxopyrrolidine-3-carboxylic acid, 1-4-chlorophenethyl-5-oxo-3-pyrrolidinecarboxylic acid, maybridge3_004495, 3-pyrrolidinecarboxylicacid, 1-2-4-chlorophenyl ethyl-5-oxo
PubChem CID 2779686
IUPAC Name 1-[2-(4-chlorophenyl)ethyl]-5-oxopyrrolidine-3-carboxylic acid
SMILES C1C(CN(C1=O)CCC2=CC=C(C=C2)Cl)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 267.71
Percent Purity 97%
Chemical Name or Material 1-(4-Chlorophenethyl)-5-oxo-3-pyrrolidinecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.