missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-1H-pyrrole-2-carbaldehyde, Tech., Thermo Scientific™

Product Code. 10783653
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10783653 1 g Amber Glass Bottle 1g
10621313 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10783653 Supplier Thermo Scientific Maybridge Supplier No. SEW04034DA
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 37560-50-2
Molecular Formula C11H8ClNO
Molecular Weight (g/mol) 205.641
MDL Number MFCD00174283
InChI Key NNQOVJLQFYROBU-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl-1h-pyrrole-2-carbaldehyde, 1-4-chlorophenyl pyrrole-2-carbaldehyde, 1-p-chlorphenyl-2-pyrrolcarbaldehyd, 1-4-chloro-phenyl-1h-pyrrole-2-carbaldehy de, 1h-pyrrole-2-carboxaldehyde,1-4-chlorophenyl
PubChem CID 737212
IUPAC Name 1-(4-chlorophenyl)pyrrole-2-carbaldehyde
SMILES C1=CN(C(=C1)C=O)C2=CC=C(C=C2)Cl

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.64
Percent Purity Tech.
Grade Technical
Chemical Name or Material 1-(4-Chlorophenyl)-1H-pyrrole-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.