missing translation for 'onlineSavingsMsg'
Learn More

1-(4-chlorophenyl)-5-methyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10467232
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber glass bottle
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10467232 1 g Amber glass bottle 1g
10148754 5 g Amber glass bottle 5g
10632253 10 g Amber glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10467232 Supplier Thermo Scientific Maybridge Supplier No. CD10529DA
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 187998-35-2
Molecular Formula C11H9ClN2O2
Molecular Weight (g/mol) 236.655
MDL Number MFCD01566260
InChI Key OAPQKGGVDCZENK-UHFFFAOYSA-N
PubChem CID 2777166
IUPAC Name 1-(4-chlorophenyl)-5-methylpyrazole-4-carboxylic acid
SMILES CC1=C(C=NN1C2=CC=C(C=C2)Cl)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym 1-4-chlorophenyl-5-methyl-1h-pyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-methylpyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-methyl-1h-pyrazole-4-carboxylicacid, 4-carboxy-1-4-chlorophenyl-5-methyl-1h-pyrazole, 1h-pyrazole-4-carboxylicacid, 1-4-chlorophenyl-5-methyl, buttpark 9618-16, salor-int l317632-1ea, 1-4-chlorophenyl-5-methylpyrazole-4-carboxylic-acid, 4-chloro-4-carboxy-5-methyl-1h-pyrazol-1-yl benzene, 1h-pyrazole-4-carboxylicacid,1-4-chlorophenyl-5-methyl
Formula Weight 236.66
Percent Purity 97%
Chemical Name or Material 1-(4-Chlorophenyl)-5-methyl-1H-pyrazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.