missing translation for 'onlineSavingsMsg'
Learn More

1-(4-chlorophenyl)-5-propyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10521872
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10521872 1 g Amber Glass Bottle 1g
10222882 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10521872 Supplier Thermo Scientific Maybridge Supplier No. GK01547DA
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175137-17-4
Molecular Formula C13H13ClN2O2
Molecular Weight (g/mol) 264.709
MDL Number MFCD00068141
InChI Key RTLVBOYPVGHHGC-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl-5-propyl-1h-pyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-propylpyrazole-4-carboxylic acid, 1h-pyrazole-4-carboxylicacid, 1-4-chlorophenyl-5-propyl, maybridge1_003964, 1h-pyrazole-4-carboxylicacid,1-4-chlorophenyl-5-propyl, 1-4-chloro-phenyl-5-propyl-1h-pyrazole-4-carboxylic acid
PubChem CID 2777583
IUPAC Name 1-(4-chlorophenyl)-5-propylpyrazole-4-carboxylic acid
SMILES CCCC1=C(C=NN1C2=CC=C(C=C2)Cl)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 264.71
Percent Purity 97%
Chemical Name or Material 1-(4-chlorophenyl)-5-propyl-1H-pyrazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.