missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10662705
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10662705 25 g Amber Glass Bottle 25g
10005962 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10662705 Supplier Thermo Scientific Maybridge Supplier No. GK01550EB
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 98534-80-6
Molecular Formula C11H6ClF3N2O2
Molecular Weight (g/mol) 290.626
MDL Number MFCD00068142
InChI Key HSZJMDBOSZVDOK-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl-5-trifluoromethyl-1h-pyrazole-4-carboxylic acid, 2-4-chlorophenyl-3-trifluoromethyl pyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-trifluoromethyl pyrazole-4-carboxylic acid, 2-4-chlorophenyl-3-trifluoromethyl-2h-pyrazole-4-carboxylic acid, maybridge1_004008, 2-4-chlorophenyl-3-trifluoromethyl pyrazole-4, 1-4-chlorophenyl-5-trifluoromethyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-trifluoromethyl
PubChem CID 688687
IUPAC Name 1-(4-chlorophenyl)-5-(trifluoromethyl)pyrazole-4-carboxylic acid
SMILES C1=CC(=CC=C1N2C(=C(C=N2)C(=O)O)C(F)(F)F)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 290.63
Percent Purity 97%
Chemical Name or Material 1-(4-Chlorophenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.