missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)biguanide hydrochloride, 97%

Product Code. 11315309
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11315309 1 g 1g
11325309 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11315309

Brand: Thermo Scientific Alfa Aesar B20311.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4022-81-5 | C8H11Cl2N5 | 248.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4022-81-5
Molecular Formula C8H11Cl2N5
Molecular Weight (g/mol) 248.11
MDL Number MFCD00053020
InChI Key NAFSLMFLGYXGIF-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl biguanide hydrochloride, 1-p-chlorophenyl biguanide hydrochloride, 1-carbamimidamido-n-4-chlorophenyl methanimidamide hydrochloride, n-p-chlorophenyl biguanide hydrochloride, 4-chloroanilino imino methyl amino methanimidamide hydrochloride, n-4-chlorophenyl-imidocarbonimidic diamide monohydrochloride, 1-carbamimidoyl-3-4-chlorophenyl guanidine, acmc-20apaw, n-p-chlorophenylbiguanide hydrochloride, 1-4-chlorophenyl-biguanide hydrochloride
PubChem CID 458746
SMILES [Cl-].NC([NH3+])=NC(N)=NC1=CC=C(Cl)C=C1

Specifications

Melting Point 247°C to 250°C
Quantity 1 g
Beilstein 3732533
IUPAC Name N-[amino(azaniumyl)methylidene]-N''-(4-chlorophenyl)guanidine chloride
Formula Weight 248.12
Percent Purity 97%
Chemical Name or Material 1-(4-Chlorophenyl)biguanide hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.