missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)cyclohexane-1-carboxylic acid, 95%

Product Code. 11347405
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347405 25 g 25g
11337405 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347405 Supplier Thermo Scientific Alfa Aesar Supplier No. L05336.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 58880-37-8 | C13H14ClO2 | 237.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58880-37-8
Molecular Formula C13H14ClO2
Molecular Weight (g/mol) 237.70
MDL Number MFCD00019350
InChI Key UPNXUJXIIZGXLQ-UHFFFAOYSA-M
Synonym 1-4-chlorophenyl cyclohexanecarboxylic acid, 1-4-chlorophenyl cyclohexane-1-carboxylic acid, 1-4-chlorophenyl-1-cyclohexanecarboxylic acid, 1-4-chlorophenyl-1-cyclohexane-carboxylic acid, cyclohexanecarboxylic acid, 1-4-chlorophenyl, acmc-1cuiq, 4-chlorophenylcyclohexanecarboxylic acid, 4-chlorophenyl cyclohexanecarboxylic acid, 1-4-chlorophenyl cyclohexanecarboxylicacid
PubChem CID 100873
IUPAC Name 1-(4-chlorophenyl)cyclohexane-1-carboxylic acid
SMILES [O-]C(=O)C1(CCCCC1)C1=CC=C(Cl)C=C1

Specifications

Melting Point 151°C to 153°C
Quantity 25 g
Beilstein 3297868
Formula Weight 238.71
Percent Purity 95%
Chemical Name or Material 1-(4-Chlorophenyl)cyclohexane-1-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.