missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)cyclopropanecarboxylic acid, 99%

Product Code. 11499400
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499400 1 g 1g
11409410 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499400 Supplier Thermo Scientific Alfa Aesar Supplier No. H50274.03
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 72934-37-3 | C10H9ClO2 | 196.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72934-37-3
Molecular Formula C10H9ClO2
Molecular Weight (g/mol) 196.63
MDL Number MFCD00001289
InChI Key YAHLWSGIQJATGG-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl cyclopropanecarboxylic acid, 1-4-chlorophenyl cyclopropane-1-carboxylic acid, 1-4-chlorophenyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-4-chlorophenyl, 1-p-chlorophenyl cyclopropanecarboxylic acid, acmc-209oos, 1-4-chlorophenyl cyclopropanecarboxylicacid, 1-4chloro-phenyl-cyclopropylcarboxylic acid, 1-4-chloro-phenyl-cyclopropylcarboxylic acid
PubChem CID 98606
IUPAC Name 1-(4-chlorophenyl)cyclopropane-1-carboxylic acid
SMILES C1CC1(C2=CC=C(C=C2)Cl)C(=O)O

Specifications

Melting Point 152°C to 155°C
Quantity 1 g
Formula Weight 196.64
Percent Purity 99%
Chemical Name or Material 1-(4-Chlorophenyl)cyclopropanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.