missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)ethylamine, 90%

Product Code. 10329420 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10329420 5 g 5g
10687943 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10329420 Supplier Thermo Scientific Acros Supplier No. 329470050
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6299-02-1 | C8H10ClN | 155.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6299-02-1
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.63
MDL Number MFCD00044766
InChI Key PINPOEWMCLFRRB-UHFFFAOYNA-N

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Yellow
Density 1.1000g/mL
Boiling Point 232.0°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula ClC6H4CH(CH3)NH2
Refractive Index 1.5414
Quantity 5 g
Specific Gravity 1.1
Synonym 1-4-chlorophenyl ethanamine, 1-4-chlorophenyl ethylamine, 4-chloro-alpha-methylbenzylamine, 1-4-chlorophenyl ethan-1-amine, 4-chloro-alpha-methyl benzylamine, 1-4-chlorophenyl-1-ethanamine, 1-4-chloro-phenyl-ethylamine, acmc-20amkz, acmc-20apje, acmc-209jma
SMILES CC(C1=CC=C(C=C1)Cl)N
IUPAC Name 1-(4-chlorophenyl)ethanamine
PubChem CID 97803
Formula Weight 155.63
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 1-(4-Chlorophenyl)ethylamine, 90%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Chloro-a-methylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.