missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)piperazine monohydrochloride, 97%, Thermo Scientific™

Product Code. 11371387
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11371387 25 g 25g
11361387 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11371387 Supplier Thermo Scientific Alfa Aesar Supplier No. L01712.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13078-12-1
Molecular Formula C10H14Cl2N2
Molecular Weight (g/mol) 233.136
MDL Number MFCD00060187
InChI Key ZHGRQBSZTVJDHU-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl piperazine hydrochloride, para-chlorophenylpiperazine hydrochloride, timtec-bb sbb003262, piperazine, 1-4-chlorophenyl-, monohydrochloride, 4-chlorophenyl piperazine, chloride, 1-4-chlorophenyl piperazine;hydrochloride, 1-4-chlorophenyl-piperazine hydrochloride, acmc-1c3h8, 1-4-chlorophenyl piperazine hcl, 1-4-chlorophenyl piperazinehydrochloride
PubChem CID 3084720
IUPAC Name 1-(4-chlorophenyl)piperazine;hydrochloride
SMILES C1CN(CCN1)C2=CC=C(C=C2)Cl.Cl

Specifications

Melting Point ∽277°C (decomposition)
Quantity 25 g
Beilstein 5647914
Formula Weight 233.14
Percent Purity 97%
Chemical Name or Material 1-(4-Chlorophenyl)piperazine monohydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.