missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexadiene, 97% stab. with 0.1% BHT

Product Code. 11381857
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381857 5 g 5g
11391857 25 g 25g
11301867 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11381857 Supplier Thermo Scientific Alfa Aesar Supplier No. L07337.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 628-41-1 | C6H8 | 80.13 g/mol

1,4-Cyclohexadiene is used to study the formation of parent ion from heavy fragmentation on irradiation with a high-intensity laser pulse. It is also used as a very effective hydrogen donor for catalytic hydrogenation reaction. It forms benzene at elevated temperatures in the presence of a ruthenium(II)-triphenylphosphine catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Cyclohexadiene is used to study the formation of parent ion from heavy fragmentation on irradiation with a high-intensity laser pulse. It is also used as a very effective hydrogen donor for catalytic hydrogenation reaction. It forms benzene at elevated temperatures in the presence of a ruthenium(II)-triphenylphosphine catalyst.

Solubility
Miscible with organic solvents like diethyl ether, tetrahydrofuran and toluene.Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-41-1
Molecular Formula C6H8
Molecular Weight (g/mol) 80.13
MDL Number MFCD00001535
InChI Key UVJHQYIOXKWHFD-UHFFFAOYSA-N
Synonym 1,4-cyclohexadiene, 1,4-dihydrobenzene, 1,4-cyclohexanediene, unii-0f8z5909qz, 1,4 cyclohexadiene, acmc-209snj, ksc354m7f, 1,4-cyclohexadiene, stabilized 5g, 1,4-cyclohexadiene, purum gc
PubChem CID 12343
ChEBI CHEBI:37611
IUPAC Name cyclohexa-1,4-diene
SMILES C1C=CCC=C1

Specifications

Melting Point -49°C
Density 0.847
Boiling Point 88°C to 90°C
Flash Point −6°C (21°F)
Refractive Index 1.472
Quantity 5 g
UN Number UN3295
Beilstein 1900733
Solubility Information Miscible with organic solvents like diethyl ether,tetrahydrofuran and toluene.Immiscible with water.
Formula Weight 80.13
Percent Purity 97%
Chemical Name or Material 1,4-Cyclohexadiene, stabilized with 0.1% BHT
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.