missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexadiene, 97%, stabilized

Product Code. 10695202 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10695202 25 mL Glass bottle 25mL
10706561 5 mL Glass bottle 5mL
10389323 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10695202 Supplier Thermo Scientific Acros Supplier No. 154820250
 Restricted Item
Estimated Shipment: 15-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 628-41-1 | C6H8 | 80.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-41-1
Molecular Formula C6H8
Molecular Weight (g/mol) 80.13
MDL Number MFCD00001535
InChI Key UVJHQYIOXKWHFD-UHFFFAOYSA-N
Synonym 1,4-cyclohexadiene, 1,4-dihydrobenzene, 1,4-cyclohexanediene, unii-0f8z5909qz, 1,4 cyclohexadiene, acmc-209snj, ksc354m7f, 1,4-cyclohexadiene, stabilized 5g, 1,4-cyclohexadiene, purum gc
PubChem CID 12343
ChEBI CHEBI:37611
IUPAC Name cyclohexa-1,4-diene
SMILES C1C=CCC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -49.2°C
Color Colorless
Density 0.8400g/mL
Boiling Point 81.0°C to 82.0°C
Flash Point −6°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4710 to 1.4730
Quantity 25 mL
Specific Gravity 0.84
Stabilizer 0.1% BHT
Formula Weight 80.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 4-Cyclohexadiene, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Cyclohexadiene, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.