missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedimethanol, Mixture of cis and trans 99%, ACROS Organics™

Product Code. 10285670 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
500g; Glass bottle
This item is not returnable. View return policy

Product Code. 10285670

Brand: Thermo Scientific Acros 154925000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 105-08-8
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H16O2
Linear Formula C6H10(CH2OH)2
Refractive Index 1.4880 to 1.4900 (20°C, 589nm)
MDL Number MFCD00001512,MFCD00066360
Quantity 500 g
Synonym 1,4-cyclohexanedimethanol, trans-1,4-cyclohexanedimethanol, chdm, 1,4-bis hydroxymethyl cyclohexane, rikabinol dm, cyclohexane-1,4-dimethanol, 1,4-chidm, 1,4-dimethylolcyclohexane, 4-hydroxymethyl cyclohexyl methanol, 1,4-cyclohexanedimethanol, cis
InChI Key YIMQCDZDWXUDCA-UHFFFAOYSA-N
SMILES OCC1CCC(CO)CC1
IUPAC Name [4-(hydroxymethyl)cyclohexyl]methanol
Molecular Weight (g/mol) 144.21
PubChem CID 7735
Formula Weight 144.21
Percent Purity ≥98.5%
Physical Form Viscous Liquid After Melting
Chemical Name or Material 1, 4-Cyclohexanedimethanol, 99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.