missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedione mono-2,2-dimethyltrimethylene ketal, 95%, Thermo Scientific™

Product Code. 11357456
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357456 25 g 25g
11337456 1 g 1g
11347456 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11357456 Supplier Thermo Scientific Alfa Aesar Supplier No. L09490.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69225-59-8
Molecular Formula C11H18O3
Molecular Weight (g/mol) 198.262
MDL Number MFCD00006652
InChI Key COKVDTKAWIFNTH-UHFFFAOYSA-N
Synonym 3,3-dimethyl-1,5-dioxaspiro 5.5 undecan-9-one, 1,4-cyclohexanedione mono-2,2-dimethyltrimethylene ketal, 1,4-cyclohexanedione mono 2,2-dimethyltrimethylene ketal, 3,3-dimethyl-1,5-dioxa-spiro 5.5 undecan-9-one, 9,9-dimethyl-7,11-dioxaspiro 5.5 undecan-3-one, 1,5-dioxaspiro 5.5 undecan-9-one, 3,3-dimethyl, pubchem11226, maybridge1_004236, 1,4-cyclohexanedionemono-2,2-dimethyltrimethyleneketal
PubChem CID 587968
IUPAC Name 3,3-dimethyl-1,5-dioxaspiro[5.5]undecan-9-one
SMILES CC1(COC2(CCC(=O)CC2)OC1)C

Specifications

Melting Point 46°C to 50°C
Quantity 25 g
Beilstein 3541930
Formula Weight 198.26
Percent Purity 95%
Chemical Name or Material 1,4-Cyclohexanedione mono-2,2-dimethyltrimethylene ketal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.