missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedione mono(2,2-dimethyltrimethylene ketal), 98+%, Thermo Scientific™

Product Code. 10192364 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10192364 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10192364 Supplier Thermo Scientific Acros Supplier No. 201520100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 69225-59-8
Melting Point 49.0°C to 50.0°C
Flash Point >110°C
Assay Percent Range 98% min. (GC)
Molecular Formula C11H18O3
Quantity 10 g
Synonym 3,3-dimethyl-1,5-dioxaspiro 5.5 undecan-9-one, 1,4-cyclohexanedione mono-2,2-dimethyltrimethylene ketal, 1,4-cyclohexanedione mono 2,2-dimethyltrimethylene ketal, 3,3-dimethyl-1,5-dioxa-spiro 5.5 undecan-9-one, 9,9-dimethyl-7,11-dioxaspiro 5.5 undecan-3-one, 1,5-dioxaspiro 5.5 undecan-9-one, 3,3-dimethyl, pubchem11226, maybridge1_004236, 1,4-cyclohexanedionemono-2,2-dimethyltrimethyleneketal
InChI Key COKVDTKAWIFNTH-UHFFFAOYSA-N
SMILES CC1(COC2(CCC(=O)CC2)OC1)C
IUPAC Name 3,3-dimethyl-1,5-dioxaspiro[5.5]undecan-9-one
Molecular Weight (g/mol) 198.26
PubChem CID 587968
Formula Weight 198.26
Percent Purity 98+%
Chemical Name or Material 1, 4-Cyclohexanedione mono(2, 2-dimethyltrimethylene ketal)
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Cyclohexanedione mono(2,2-dimethyltrimethylene ketal)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.