missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Cyclohexanedione Monoethylene Acetal, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 1,4-Cyclohexanedione Monoethylene Acetal |
| CAS | 4746-97-8 |
| Molecular Formula | C8H12O3 |
| Quantity | 25 g |
| Synonym | 1,4-Dioxaspiro[4.5]decan-8-one, 1,4-Cyclohexanedione Cyclic Ethylene Monoketal, 1,4-Cyclohexanedione Mono(ethylene Ketal), 1,4-Cyclohexanedione Monoethylene Ketal, 1,4-Cyclohexanedione Cyclic 1,2-Ethanediyl Acetal, 4,4-(Ethylenedioxy)cyclohexanone, 6,10-Dioxaspiro[4.5]decan-8-one, 8-Oxo-1,4-dioxaspiro[4.5]decane, Cyclohexane-1,4-dione Mono(ethylene Glycol Ketal) |
| InChI Formula | InChI=1S/C8H12O3/c9-7-1-3-8(4-2-7)10-5-6-11-8/h1-6H2 |
| SMILES | O=C1CCC2(CC1)OCCO2 |
| IUPAC Name | 1,4-dioxaspiro[4.5]decan-8-one |
| Molecular Weight (g/mol) | 156.18 |
| Formula Weight | 156.0786 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction