missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedione Monoethylene Acetal, TRC

Product Code. 30350106
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30350106

Brand: Toronto Research Chemicals TRCC9879555G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,4-Cyclohexanedione Monoethylene Acetal
CAS 4746-97-8
Molecular Formula C8H12O3
Quantity 5 g
Synonym 1,4-Dioxaspiro[4.5]decan-8-one, 1,4-Cyclohexanedione Cyclic Ethylene Monoketal, 1,4-Cyclohexanedione Mono(ethylene Ketal), 1,4-Cyclohexanedione Monoethylene Ketal, 1,4-Cyclohexanedione Cyclic 1,2-Ethanediyl Acetal, 4,4-(Ethylenedioxy)cyclohexanone, 6,10-Dioxaspiro[4.5]decan-8-one, 8-Oxo-1,4-dioxaspiro[4.5]decane, Cyclohexane-1,4-dione Mono(ethylene Glycol Ketal)
InChI Formula InChI=1S/C8H12O3/c9-7-1-3-8(4-2-7)10-5-6-11-8/h1-6H2
SMILES O=C1CCC2(CC1)OCCO2
IUPAC Name 1,4-dioxaspiro[4.5]decan-8-one
Molecular Weight (g/mol) 156.18
Formula Weight 156.0786
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.