missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Cyclohexanedione, TRC
Click to view available options
Quantity:
25 g
Specifications
Specifications
| Chemical Name or Material | 1,4-Cyclohexanedione |
| CAS | 637-88-7 |
| Molecular Formula | C6H8O2 |
| Quantity | 25 g |
| Synonym | 1,4-Dioxocyclohexane, NSC 7192, Tetrahydroquinone |
| InChI Formula | InChI=1S/C6H8O2/c7-5-1-2-6(8)4-3-5/h1-4H2 |
| SMILES | O=C1CCC(=O)CC1 |
| IUPAC Name | cyclohexane-1,4-dione |
| Molecular Weight (g/mol) | 112.13 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction