missing translation for 'onlineSavingsMsg'
Learn More

1,4-Di-n-butylbenzene, 97+%

Product Code. 15489029
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 15489029

Brand: Thermo Scientific Alfa Aesar L16396.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1571-86-4 | C14H22 | 190.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1571-86-4
Molecular Formula C14H22
Molecular Weight (g/mol) 190.33
MDL Number MFCD01076591
InChI Key VGQOZYOOFXEGDA-UHFFFAOYSA-N
Synonym 1,4-di-n-butylbenzene, benzene, 1,4-dibutyl, p-dibutylbenzol, p-dibutylbenzene, acmc-209dfm, p-di-tert-butyl benzene
PubChem CID 519170
IUPAC Name 1,4-dibutylbenzene
SMILES CCCCC1=CC=C(CCCC)C=C1

Specifications

Melting Point -31°C
Density 0.856
Boiling Point 227°C
Flash Point 110°C (230°F)
Refractive Index 1.489
Quantity 1 g
Beilstein 1858055
Formula Weight 190.33
Percent Purity ≥97%
Chemical Name or Material 1,4-Di-n-butylbenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.