missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diaminobutane dihydrochloride, 99+%

Product Code. 10175220 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10175220 25 g Glass bottle 25g
10430681 5 g Glass bottle 5g
10229453 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10175220 Supplier Thermo Scientific Acros Supplier No. 215110250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 333-93-7 | C4H12N2·2HCl | 161.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-93-7
Molecular Formula C4H12N2·2HCl
Molecular Weight (g/mol) 161.08
MDL Number MFCD00012526
InChI Key XXWCODXIQWIHQN-UHFFFAOYSA-N
Synonym 1,4-diaminobutane dihydrochloride, putrescine dihydrochloride, tetramethylenediamine dihydrochloride, putrescine hydrochloride, 1,4-butanediamine dihydrochloride, unii-x45sur7rhy, 1,4-butanediamine, dihydrochloride, 1,4-butanediamine, hydrochloride, 1,4-diaminobutane, dihydrochloride, butane-1,4-diamine dihydrochloride
PubChem CID 9532
IUPAC Name butane-1,4-diamine;dihydrochloride
SMILES C(CCN)CN.Cl.Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 280.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula H2N(CH2)4NH2·2HCl
Quantity 25 g
Beilstein 04, 264
Merck Index 15, 8058
Solubility Information Solubility in water: soluble.
Formula Weight 161.08
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 1, 4-Diaminobutane dihydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Diaminobutane dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.