missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diaminobutane dihydrochloride, 99%

Product Code. 11453867
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453867 25 g 25g
11463867 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11453867 Supplier Thermo Scientific Alfa Aesar Supplier No. A18312.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 333-93-7 | C4H14Cl2N2 | 161.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-93-7
Molecular Formula C4H14Cl2N2
Molecular Weight (g/mol) 161.07
MDL Number MFCD00012526
InChI Key XXWCODXIQWIHQN-UHFFFAOYSA-N
Synonym 1,4-diaminobutane dihydrochloride, putrescine dihydrochloride, tetramethylenediamine dihydrochloride, putrescine hydrochloride, 1,4-butanediamine dihydrochloride, unii-x45sur7rhy, 1,4-butanediamine, dihydrochloride, 1,4-butanediamine, hydrochloride, 1,4-diaminobutane, dihydrochloride, butane-1,4-diamine dihydrochloride
PubChem CID 9532
IUPAC Name butane-1,4-diamine;dihydrochloride
SMILES C(CCN)CN.Cl.Cl

Specifications

Melting Point ∼280°C (decomposition)
Quantity 25 g
Beilstein 3906680
Sensitivity Hygroscopic
Merck Index 14,7947
Formula Weight 161.08
Percent Purity 99%
Chemical Name or Material 1,4-Diaminobutane dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.