missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diaminobutane, TRC

Product Code. 30135224
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135224 100 g 100g
30136904 25 g 25g
30005187 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135224 Supplier Toronto Research Chemicals Supplier No. TRCD416025100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-60-1
Formula Weight 88.1
InChI Formula InChI=1S/C4H12N2/c5-3-1-2-4-6/h1-6H2
IUPAC Name butane-1,4-diamine
Molecular Formula C4 H12 N2
Molecular Weight (g/mol) 88.15
SMILES NCCCCN
Synonym Tetramethylenediamine (7CI), 1,4-Butylenediamine, 1,4-Diamino-n-butane, 1,4-Diaminobutane, 1,4-Tetramethylenediamine, NSC 60545, Putramine, Putrascine, Putrescin, Putrescine, α,ω-Butanediamine
Chemical Name or Material 1,4-Diaminobutane

Specifications

Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.