missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diazabicyclo[2.2.2]octane bis(Sulfur Dioxide) Adduct, TRC

Product Code. 30353396
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353396 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30353396 Supplier Toronto Research Chemicals Supplier No. TRCD41760350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,4-Diazabicyclo[2.2.2]octane bis(Sulfur Dioxide) Adduct
CAS 119752-83-9
Molecular Formula C6H12N2O4S2
Quantity 50 mg
Synonym 1,4-Disulfino-1,4-Diazoniabicyclo[2.2.2]Octane, Bis(Inner Salt)
InChI Formula InChI=1S/C6H12N2O4S2/c9-13(10)7-1-2-8(5-3-7,6-4-7)14(11)12/h1-6H2
SMILES O=[S-](=O)[N+]12CC[N+](CC1)(CC2)[S-](=O)=O
IUPAC Name 1,4-disulfinato-1,4-diazoniabicyclo[2.2.2]octane
Molecular Weight (g/mol) 240.3
Formula Weight 240.0238
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.