missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diazepane, 97%, Thermo Scientific™

Product Code. 10157384
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10157384 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10157384 Supplier Thermo Scientific Maybridge Supplier No. SB01866DE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 505-66-8
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
InChI Key FQUYSHZXSKYCSY-UHFFFAOYSA-N
Synonym homopiperazine, 1,4-diazacycloheptane, 1h-1,4-diazepine, hexahydro, hexahydro-1,4-diazepine, 1,4 diazepane, perhydro-1,4-diazepine, trimethyleneethylenediamine, 1,4-diazepan, unii-95cl167w8t, hexahydro-1h-1,4-diazepine
PubChem CID 68163
IUPAC Name 1,4-diazepane
SMILES C1CNCCNC1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 100.16
Percent Purity 95%
Chemical Name or Material 1, 4-Diazepane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.